1-(Benzyloxy)-5-bromo-3-chloro-2-fluorobenzene

Supplier: AOBChem USA

Warning

72803-500MG 72803-1G 72803-5G 72803-25G
77333-832EA 194.67 USD
77333-832 77357-994 77369-846 77390-740
1-(Benzyloxy)-5-bromo-3-chloro-2-fluorobenzene
1-(Benzyloxy)-5-bromo-3-chloro-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉BrClFO
MW: 315.565 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29043958
CAS Number: 1881320-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR