1-Ethoxy-2-fluoro-3-iodo-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

72794-500MG 72794-1G 72794-5G 72794-25G
77333-830EA 116.39 USD
77333-830 77343-464 77369-844 77381-906
1-Ethoxy-2-fluoro-3-iodo-4-methoxybenzene ≥97%
1-Ethoxy-2-fluoro-3-iodo-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀FIO₂
MW: 296.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764560
CAS Number: 2385093-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR