(3-Ethoxy-2-fluoro-6-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

72793-500MG 72793-1G 72793-5G 72793-25G
77333-828EA 101.97 USD
77333-828 77343-016 77369-842 77381-904
(3-Ethoxy-2-fluoro-6-methoxyphenyl)(methyl)sulfane ≥97%
(3-Ethoxy-2-fluoro-6-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO₂S
Storage Temperature: Ambient
MDL Number: MFCD34764559
CAS Number: 2711246-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR