3-Ethoxy-2-fluoro-6-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

72789-1G 72789-5G 72789-25G
77341-606EA 94.76 USD
77341-606 77369-840 77381-902
3-Ethoxy-2-fluoro-6-methoxybenzoic acid ≥97%
3-Ethoxy-2-fluoro-6-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₄
MW: 214.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764555
CAS Number: 2382873-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR