3,6-Diethoxy-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

72780-500MG 72780-1G 72780-5G 72780-25G
77333-822EA 182.31 USD
77333-822 77347-026 77369-832 77390-738
3,6-Diethoxy-2-fluorobenzoic acid ≥97%
3,6-Diethoxy-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₄
MW: 228.217 g/mol
Storage Temperature: Ambient
CAS Number: 2918965-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR