(3,6-Diethoxy-2-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

72777-500MG 72777-1G 72777-5G 72777-25G
77333-820EA 197.76 USD
77333-820 77357-988 77369-830 77390-736
(3,6-Diethoxy-2-fluorophenyl)(methyl)sulfane ≥97%
(3,6-Diethoxy-2-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₁H₁₅FO₂S
MW: 230.299 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764544
CAS Number: 2807470-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR