1,4-Diethoxy-2-fluoro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

72776-500MG 72776-1G 72776-5G 72776-25G
77333-818EA 180.25 USD
77333-818 77346-768 77369-828 77390-734
1,4-Diethoxy-2-fluoro-3-iodobenzene ≥97%
1,4-Diethoxy-2-fluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂FIO₂
Storage Temperature: Ambient
MDL Number: MFCD34764543
CAS Number: 2807450-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR