1-Butoxy-2-fluoro-4-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

72773-500MG 72773-1G 72773-5G 72773-25G
77333-816EA 120.51 USD
77333-816 77343-820 77369-826 77381-894
1-Butoxy-2-fluoro-4-iodobenzene ≥97%
1-Butoxy-2-fluoro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂FIO
MW: 294.105 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764540
CAS Number: 2807447-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR