4-Bromo-1-butoxy-2-methoxybenzene

Supplier: AOBChem USA

Warning

72771-500MG 72771-1G 72771-5G 72771-25G
77333-812EA 163.77 USD
77333-812 77346-130 77369-822 77390-730
4-Bromo-1-butoxy-2-methoxybenzene
4-Bromo-1-butoxy-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO₂
MW: 259.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917830
CAS Number: 330475-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR