1-Bromo-2,4-diethoxy-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

72761-500MG 72761-1G 72761-5G
77335-912EA 367.71 USD
77335-912 77357-984 77381-888
1-Bromo-2,4-diethoxy-3-fluorobenzene ≥97%
1-Bromo-2,4-diethoxy-3-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO₂
MW: 263.103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764530
CAS Number: 2807466-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR