2-(Benzyloxy)-1-bromo-4-ethoxy-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

72760-500MG 72760-1G 72760-5G
77336-570EA 388.31 USD
77336-570 77357-982 77381-886
2-(Benzyloxy)-1-bromo-4-ethoxy-3-fluorobenzene ≥97%
2-(Benzyloxy)-1-bromo-4-ethoxy-3-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34764529
CAS Number: 2807463-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR