(4-Fluoro-3-isopropoxy-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

72754-500MG 72754-1G 72754-5G 72754-25G
77333-800EA 148.32 USD
77333-800 77345-284 77369-814 77390-724
(4-Fluoro-3-isopropoxy-5-methylphenyl)(methyl)sulfane ≥97%
(4-Fluoro-3-isopropoxy-5-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₁H₁₅FOS
MW: 214.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764523
CAS Number: 2807445-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR