2-Fluoro-5-iodo-1-isopropoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

72753-500MG 72753-1G 72753-25G 72753-5G
77333-798EA 291.49 USD
77333-798 77357-978 77390-722 77381-882
2-Fluoro-5-iodo-1-isopropoxy-3-methylbenzene ≥97%
2-Fluoro-5-iodo-1-isopropoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂FIO
Storage Temperature: Ambient
MDL Number: MFCD34764522
CAS Number: 2807444-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR