1-(Difluoromethoxy)-2-fluoro-5-iodo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

72752-500MG 72752-1G 72752-5G
77335-454EA 353.29 USD
77335-454 77357-976 77381-880
1-(Difluoromethoxy)-2-fluoro-5-iodo-3-methylbenzene ≥97%
1-(Difluoromethoxy)-2-fluoro-5-iodo-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₆F₃IO
MW: 302.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764521
CAS Number: 2386338-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR