1,3,5-Tribromo-2-chlorobenzene ≥97%

Supplier: AOBChem USA

Warning

72741-500MG 72741-1G 72741-5G
77338-928EA 531.48 USD
77338-928 77357-974 77381-878
1,3,5-Tribromo-2-chlorobenzene ≥97%
1,3,5-Tribromo-2-chlorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₃Cl
MW: 349.245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18400035
CAS Number: 78904-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR