1-Chloro-2-(difluoromethoxy)-5-fluoro-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

72740-500MG 72740-1G 72740-5G
77338-816EA 519.12 USD
77338-816 77357-972 77381-876
1-Chloro-2-(difluoromethoxy)-5-fluoro-3-methylbenzene ≥95%
1-Chloro-2-(difluoromethoxy)-5-fluoro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₆ClF₃O
MW: 210.581 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28750737
CAS Number: 1807240-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR