2,3-Dibromo-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

72730-500MG 72730-1G 72730-5G
77339-812EA 649.93 USD
77339-812 77357-968 77381-874
2,3-Dibromo-5-methoxybenzoic acid ≥97%
2,3-Dibromo-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₃
MW: 309.939 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764510
CAS Number: 2092356-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR