(4-Methoxy-2-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

72723-500MG 72723-1G 72723-5G 72723-25G
77333-792EA 252.35 USD
77333-792 77357-964 77369-808 77390-716
(4-Methoxy-2-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(4-Methoxy-2-methylphenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD29070834
CAS Number: 103562-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR