(4-Methoxy-2-methylphenyl)(piperidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

72720-500MG 72720-1G 72720-5G 72720-25G
77333-788EA 260.59 USD
77333-788 77357-962 77369-804 77390-712
(4-Methoxy-2-methylphenyl)(piperidin-1-yl)methanone ≥97%
(4-Methoxy-2-methylphenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₂
MW: 233.306 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764505
CAS Number: 2807471-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR