2-(Benzyloxy)-4-bromo-1-chloro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

72693-500MG 72693-1G 72693-5G 72693-25G
77333-780EA 110.21 USD
77333-780 77343-280 77369-798 77381-870
2-(Benzyloxy)-4-bromo-1-chloro-3-methylbenzene ≥97%
2-(Benzyloxy)-4-bromo-1-chloro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrClO
MW: 311.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764494
CAS Number: 2901065-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR