1-Bromo-4-chloro-3-isopropoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

72692-500MG 72692-1G 72692-5G 72692-25G
77333-778EA 183.34 USD
77333-778 77347-136 77369-796 77390-706
1-Bromo-4-chloro-3-isopropoxy-2-methylbenzene ≥97%
1-Bromo-4-chloro-3-isopropoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrClO
MW: 263.559 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764493
CAS Number: 2901109-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR