4-(Benzyloxy)-5-fluoro-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

72665-500MG 72665-1G 72665-5G
77337-226EA 408.91 USD
77337-226 77357-952 77381-862
4-(Benzyloxy)-5-fluoro-2-methoxybenzoic acid ≥97%
4-(Benzyloxy)-5-fluoro-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₄
Storage Temperature: Ambient
MDL Number: MFCD34764468
CAS Number: 949119-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR