4-(Benzyloxy)-5-fluoro-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

72663-500MG 72663-1G 72663-5G
77338-026EA 459.38 USD
77338-026 77357-950 77381-860
4-(Benzyloxy)-5-fluoro-2-methoxybenzaldehyde ≥97%
4-(Benzyloxy)-5-fluoro-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₃
MW: 260.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764466
CAS Number: 2807441-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR