5-Fluoro-4-isopropoxy-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

72658-500MG 72658-1G 72658-5G
77336-284EA 384.19 USD
77336-284 77357-948 77381-858
5-Fluoro-4-isopropoxy-2-methoxybenzoic acid ≥97%
5-Fluoro-4-isopropoxy-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₄
MW: 228.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764461
CAS Number: 2807471-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR