5-Fluoro-4-isopropoxy-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

72656-500MG 72656-1G 72656-5G
77337-224EA 408.91 USD
77337-224 77357-946 77381-856
5-Fluoro-4-isopropoxy-2-methoxybenzaldehyde ≥97%
5-Fluoro-4-isopropoxy-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764459
CAS Number: 2807450-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR