2,2,2-Trifluoro-1-(4-propoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

72625-1G 72625-5G 72625-25G
77341-580EA 93.73 USD
77341-580 77369-794 77381-854
2,2,2-Trifluoro-1-(4-propoxyphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(4-propoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD11556211
CAS Number: 1256467-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR