2,2,2-Trifluoro-2',4',6'-trimethylacetophenone ≥97%

Supplier: AOBChem USA

Warning

72623-500MG 72623-1G 72623-5G 72623-25G
77333-776EA 120.51 USD
77333-776 77343-818 77369-792 77381-852
2,2,2-Trifluoro-2',4',6'-trimethylacetophenone ≥97%
2,2,2-Trifluoro-2',4',6'-trimethylacetophenone

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃O
MW: 216.19g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151856
CAS Number: 313-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR