2'-Isopropyl-2,2,2-trifluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,2,2-Trifluoro-1-(2-isopropylphenyl)ethanone

72621-500MG 72621-1G 72621-5G 72621-25G
77333-774EA 207.03 USD
77333-774 77357-944 77369-790 77390-704
2'-Isopropyl-2,2,2-trifluoroacetophenone ≥97%
2'-Isopropyl-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃O
MW: 216.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01319977
CAS Number: 845823-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR