1-Chloro-4-ethoxy-2-fluoro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

72611-500MG 72611-1G 72611-5G 72611-25G
77333-764EA 130.81 USD
77333-764 77344-356 77369-780 77381-846
1-Chloro-4-ethoxy-2-fluoro-3-iodobenzene ≥97%
1-Chloro-4-ethoxy-2-fluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₇ClFIO
MW: 300.496 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764445
CAS Number: 2190522-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR