3-Iodo-4-methyl-5-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

72544-500MG 72544-1G 72544-5G
77339-788EA 638.6 USD
77339-788 77357-940 77381-828
3-Iodo-4-methyl-5-(methylthio)pyridine ≥97%
3-Iodo-4-methyl-5-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₈INS
MW: 265.115 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30539994
CAS Number: 2322966-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR