2-Chloro-5-fluoro-1-iodo-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

72543-500MG 72543-1G 72543-5G 72543-25G
77333-756EA 145.23 USD
77333-756 77345-110 77369-764 77390-698
2-Chloro-5-fluoro-1-iodo-3-methoxybenzene ≥97%
2-Chloro-5-fluoro-1-iodo-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFIO
MW: 286.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764407
CAS Number: 2385419-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR