2-Chloro-5-fluoro-3-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

72539-500MG 72539-1G 72539-5G 72539-25G
77333-752EA 190.55 USD
77333-752 77357-938 77369-760 77390-696
2-Chloro-5-fluoro-3-methoxybenzoic acid ≥97%
2-Chloro-5-fluoro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₃
MW: 204.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28644344
CAS Number: 1782016-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR