1,5-Dichloro-3-methoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

72538-500MG 72538-1G
77340-454EA 1119.61 USD
77340-454 77357-936
1,5-Dichloro-3-methoxy-2-methylbenzene ≥97%
1,5-Dichloro-3-methoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈Cl₂O
MW: 191.055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786749
CAS Number: 67365-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR