(4'-Bromo-[1,1'-biphenyl]-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

72524-500MG 72524-1G 72524-5G
77337-630EA 431.57 USD
77337-630 77357-934 77381-824
(4'-Bromo-[1,1'-biphenyl]-3-yl)methanol ≥97%
(4'-Bromo-[1,1'-biphenyl]-3-yl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrO
MW: 263.1298 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05981699
CAS Number: 2002045-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR