2-Fluoro-4-iodo-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

72492-500MG 72492-1G 72492-5G 72492-25G
77333-746EA 204.97 USD
77333-746 77357-930 77369-754 77390-690
2-Fluoro-4-iodo-1,1'-biphenyl ≥97%
2-Fluoro-4-iodo-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈FI
MW: 298.095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14701835
CAS Number: 56985-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR