(4-Bromo-3-fluoro-2-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

72479-500MG 72479-1G 72479-5G 72479-25G
77333-738EA 185.4 USD
77333-738 77347-370 77369-748 77390-684
(4-Bromo-3-fluoro-2-iodophenyl)(methyl)sulfane ≥97%
(4-Bromo-3-fluoro-2-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅BrFIS
MW: 346.986 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764383
CAS Number: 2705556-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR