4-Cyclopentyl-3-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

72448-500MG 72448-1G 72448-5G
77339-526EA 594.31 USD
77339-526 77357-926 77381-818
4-Cyclopentyl-3-fluorobenzoic acid ≥97%
4-Cyclopentyl-3-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD24481490
CAS Number: 801303-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR