3,5-Dichloro-2-fluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

72438-500MG 72438-1G 72438-25G 72438-5G
77333-736EA 291.49 USD
77333-736 77357-924 77390-682 77381-816
3,5-Dichloro-2-fluoro-1,1'-biphenyl ≥97%
3,5-Dichloro-2-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇Cl₂F
Storage Temperature: Ambient
MDL Number: MFCD34764361
CAS Number: 2901086-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR