3,5-Dichloro-2-fluoro-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

72437-500MG 72437-1G 72437-5G 72437-25G
77333-734EA 251.32 USD
77333-734 77357-922 77369-746 77390-680
3,5-Dichloro-2-fluoro-4'-methyl-1,1'-biphenyl ≥97%
3,5-Dichloro-2-fluoro-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉Cl₂F
Storage Temperature: Ambient
MDL Number: MFCD34764360
CAS Number: 1392440-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR