3,5-Dichloro-2,4'-difluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

72434-500MG 72434-1G 72434-5G 72434-25G
77333-730EA 221.45 USD
77333-730 77357-918 77369-742 77390-676
3,5-Dichloro-2,4'-difluoro-1,1'-biphenyl ≥97%
3,5-Dichloro-2,4'-difluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₆Cl₂F₂
Storage Temperature: Ambient
MDL Number: MFCD34764358
CAS Number: 2901082-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR