2,3-Dichloro-6-iodophenol ≥97%

Supplier: AOBChem USA

Warning

72430-500MG 72430-1G 72430-5G 72430-25G
77333-728EA 122.57 USD
77333-728 77343-968 77369-740 77381-814
2,3-Dichloro-6-iodophenol ≥97%
2,3-Dichloro-6-iodophenol

Precaución: for Research Use Only

Formula: C₆H₃Cl₂IO
MW: 288.898 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786685
CAS Number: 1803809-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR