2,3-Dichloro-6-(methylthio)phenol ≥95%

Supplier: AOBChem USA

Warning

72428-500MG 72428-1G 72428-5G 72428-25G
77333-726EA 202.91 USD
77333-726 77357-916 77369-738 77390-674
2,3-Dichloro-6-(methylthio)phenol ≥95%
2,3-Dichloro-6-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₇H₆Cl₂OS
MW: 209.093 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786653
CAS Number: 1806270-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR