3,4-Dichloro-2-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

72423-500MG 72423-1G 72423-5G 72423-25G
77333-724EA 224.54 USD
77333-724 77357-914 77369-736 77390-672
3,4-Dichloro-2-hydroxybenzoic acid ≥97%
3,4-Dichloro-2-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄Cl₂O₃
MW: 207.011 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11203614
CAS Number: 14010-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR