4-(Benzyloxy)-1-iodo-2-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

72375-500MG 72375-1G 72375-5G 72375-25G
77333-720EA 272.95 USD
77333-720 77357-910 77369-730 77390-668
4-(Benzyloxy)-1-iodo-2-(trifluoromethyl)benzene ≥97%
4-(Benzyloxy)-1-iodo-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₃IO
MW: 378.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764344
CAS Number: 2807444-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR