4-Chloro-5-methoxy-2-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

72370-500MG 72370-1G
77340-524EA 1438.91 USD
77340-524 77357-906
4-Chloro-5-methoxy-2-(trifluoromethyl)benzoic acid ≥97%
4-Chloro-5-methoxy-2-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34764339
CAS Number: 2383858-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR