4-Chloro-5-methoxy-2-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

72368-500MG 72368-1G 72368-5G
77340-294EA 890.95 USD
77340-294 77357-904 77381-808
4-Chloro-5-methoxy-2-(trifluoromethyl)benzaldehyde ≥97%
4-Chloro-5-methoxy-2-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O₂
MW: 238.591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764337
CAS Number: 2385487-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR