2-Chloro-5-fluoro-3-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

72363-500MG 72363-1G 72363-5G
77336-568EA 388.31 USD
77336-568 77357-900 77381-804
2-Chloro-5-fluoro-3-formylbenzoic acid ≥97%
2-Chloro-5-fluoro-3-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClFO₃
MW: 202.567 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764334
CAS Number: 2384449-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR