2-Chloro-3-ethoxy-5-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

72348-500MG 72348-1G 72348-5G
77339-524EA 594.31 USD
77339-524 77357-892 77381-796
2-Chloro-3-ethoxy-5-fluorobenzaldehyde ≥97%
2-Chloro-3-ethoxy-5-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD34764319
CAS Number: 2386261-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR