2-Chloro-5-fluoro-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

72346-500MG 72346-1G 72346-5G
77339-136EA 553.11 USD
77339-136 77357-890 77381-794
2-Chloro-5-fluoro-3-methoxybenzaldehyde ≥97%
2-Chloro-5-fluoro-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34619885
CAS Number: 1896494-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR