(2-Chloro-5-fluoro-3-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

72343-500MG 72343-1G 72343-5G 72343-25G
77333-718EA 163.77 USD
77333-718 77346-128 77369-728 77390-666
(2-Chloro-5-fluoro-3-methylphenyl)(methyl)sulfane ≥97%
(2-Chloro-5-fluoro-3-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈ClFS
Storage Temperature: Ambient
MDL Number: MFCD28752228
CAS Number: 1805114-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR